Lomustine Related Compound B
Synonym(s):
1-(2-Chloroethyl)-3-cyclohexylurea, N-(2-Chloroethyl)-N′-cyclohexylurea
Empirical Formula (Hill Notation):
C9H17ClN2O
CAS Number:
Molecular Weight:
204.70
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing