Skip to Content
Merck
All Photos(1)

Documents

1123146

USP

Chlorpheniramine Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-(4-Chlorophenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN2 · C4H4O4
CAS Number:
Molecular Weight:
376.83
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorpheniramine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

MGNXGZAZRYTQMW-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Central nervous system research: Chlorpheniramine Related Compound C is examined for its effects on the central nervous system, specifically regarding histamine-related responses, providing insights for antihistamine development and neurological research (Kamei, 2021).

  • Virucidal research applications: The virucidal effects of Chlorpheniramine Related Compound C against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) are studied, demonstrating potential applications in pandemic response and viral research (Westover et al., 2020).

  • Bitter taste receptor research: Investigations into the effects of Chlorpheniramine Related Compound C on the expression of bitter taste receptor T2R7 and its signaling effectors in cellular models contribute to the understanding of taste perception and receptor activation mechanisms (Su et al., 2019).

  • Synthesis and evaluation of antihistaminic agents: Chlorpheniramine Related Compound C plays a role in the design and synthesis of new chemical entities aiming for improved antihistaminic and antiasthmatic properties, enhancing pharmaceutical innovations and therapeutic strategies (Sirisha et al., 2014).

  • Mast cell degranulation and histamine release: The compound′s involvement in studying the biological mechanisms of pain and edema induced by venom illustrates its utility in pharmacological and toxicological research, highlighting its role in elucidating histamine pathways (Liu et al., 2007).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service