Skip to Content
Merck
Chlorpheniramine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material

Chlorpheniramine Related Compound C

Chlorpheniramine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-(4-Chlorophenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine maleate salt
Empirical Formula (Hill Notation):
C15H17ClN2 · C4H4O4
CAS Number:
Molecular Weight:
376.83

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1