Skip to Content
Merck
All Photos(1)

Key Documents

1104210

USP

Cevimeline Hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(5R)-2-methylspiro[1,3-oxathiolane-5,3′-1-azabicyclo[2.2.2]octane]; hydrate; dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NOS · HCl · 0.5 H2O
CAS Number:
Molecular Weight:
244.78
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Agency

USP

API family

cevimeline

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

SMILES string

S4C(O[C@@]5(C6CCN(C5)CC6)C4)C.S1C(O[C@@]2(C3CCN(C2)CC3)C1)C.Cl.Cl.O

InChI

1S/2C10H17NOS.2ClH.H2O/c2*1-8-12-10(7-13-8)6-11-4-2-9(10)3-5-11;;;/h2*8-9H,2-7H2,1H3;2*1H;1H2/t2*8?,10-;;;/m11.../s1

InChI key

ZSTLCHCDLIUXJE-MMPAUJBDSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Cevimeline Hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service