Skip to Content
Merck
All Photos(1)

Documents

W350818

Sigma-Aldrich

Methyl 3-hydroxyhexanoate

≥97%, FG

Synonym(s):

Methyl 3-hydroxycaproate, Pineapple hydroxyhexanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CO2CH3
CAS Number:
Molecular Weight:
146.18
FEMA Number:
3508
EC Number:
Council of Europe no.:
10812
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.532
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.43 (lit.)

density

1 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

oily; woody; ethereal; fruity; pineapple; sweet

SMILES string

CCCC(O)CC(=O)OC

InChI

1S/C7H14O3/c1-3-4-6(8)5-7(9)10-2/h6,8H,3-5H2,1-2H3

InChI key

ACCRBMDJCPPJDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service