Skip to Content
Merck
All Photos(1)

Documents

816049

TSKgel® QC-PAK GFC 300 HPLC Column

phase diol, L × I.D. 15 cm × 7.8 mm, 5 μm particle size

Synonym(s):

TSKgel® SWXL Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 16049

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature
41 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 7.8 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

5 μm

pore size

250 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The TSKgel QC-PAK columns are available within the TSKgel G2000SWxl and G3000SWxl line. These columns are 15cm in length with 5μm particles and offer the same resolution in half the time as the 30cm, 10μm TSKgel G2000SW and G3000SW columns.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service