Skip to Content
Merck
All Photos(1)

Documents

808542

TSKgel® G4000SWXL (8 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, diol phase

Synonym(s):

TSKgel® SWXL Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® G4000SWXL HPLC Column, phase diol, L × I.D. 30 cm × 7.8 mm, 8 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 08542

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature
35 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

8 μm

pore size

450 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Widely considered the premium columns for analyzing biopolymers by GFC, TSKgel SWXL columns feature high pore volume per unit column volume, low sample adsorption and excellent column efficiency, all contributing to unsurpassed sample resolution. With 8μm particles and 450Å pores, TSKgel G4000SWXL provides the largest exclusion limit and the widest fractionation range. It is an excellent choice for pegylated proteins or glycosylated biomolecules. The fractionation range is applicable for globular protein samples up to 7 million Da.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service