57249-U
IonSense® PDMS/DVB SPE-it Tips
pkg of 96 ea
Synonym(s):
SPME LC TIps
About This Item
Recommended Products
packaging
pkg of 96 ea
fiber L
1 cm
matrix active group
PDMS/DVB
application(s)
food and beverages
compatibility
for use with pipettor or robotic liquid handling systems
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
For extraction of a wider range of polar and nonpolar analytes, try our SPME LC Tips, C18 (P/N 57234-U).
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
A demonstration of the use of innovative BioSPME fibers for the selective extraction of drugs and polar metbolites from biological matrices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service