Skip to Content
Merck
All Photos(4)

Documents

A5006

Sigma-Aldrich

L-Arginine

≥98%

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Arginine, reagent grade, ≥98%

grade

reagent grade

Quality Level

Assay

≥98%

form

powder

color

white

mp

222 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

application(s)

cell analysis
peptide synthesis

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-arginine has been used to study non-enzymatic gluconeogenesis. It has also been used to study the effects of L-arginine supplementation on kidney and liver injury in rats with myocardial infarction.

Biochem/physiol Actions

L-Arginine is a dibasic, semi-essential amino acid. It acts as a precursor for creatinine and is a natural constituent of most of the dietary proteins.
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.P.F. D'Mello
Amino Acids in Human Nutrition and Health (2012)
Aerobic training and L-arginine supplement attenuates myocardial infarction-induced kidney and liver injury in rats via reduces oxidative stress
Kamal Ranjbar
Indian Heart Journal (2017)
Nonenzymatic gluconeogenesis-like formation of
fructose 1,6-bisphosphate in ice
Christoph B. Messner
Proceedings of the National Academy of Sciences of the USA, 7403-7407 (2017)
Krishnan Suresh Kumar et al.
European journal of medicinal chemistry, 45(11), 5474-5479 (2010-08-21)
A new series of 3-(benzylideneamino)-2-phenylquinazoline-4(3H)-ones were prepared through Schiff base formation of 3-amino-2-phenyl quinazoline-4(3)H-one with various substituted carbonyl compounds. Their chemical structures were elucidated by spectral studies. Cytotoxicity and antiviral activity were evaluated against herpes simplex virus-1 (KOS), herpes simplex
Tomas Del Olmo et al.
EMBO reports, 20(2) (2019-01-06)
RAB GTPases are central modulators of membrane trafficking. They are under the dynamic regulation of activating guanine exchange factors (GEFs) and inactivating GTPase-activating proteins (GAPs). Once activated, RABs recruit a large spectrum of effectors to control trafficking functions of eukaryotic

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service