Skip to Content
Merck
All Photos(1)

Key Documents

442784

Supelco

Squalane

analytical standard

Synonym(s):

2,6,10,15,19,23-Hexamethyltetracosane, Cosbiol, Perhydrosqualene, Robane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)CH2CH2-]2
CAS Number:
Molecular Weight:
422.81
Beilstein:
776019
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.452 (lit.)

bp

176 °C/0.05 mmHg (lit.)

mp

−38 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages

format

neat

storage temp.

room temp

SMILES string

CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

InChI

1S/C30H62/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h25-30H,9-24H2,1-8H3

InChI key

PRAKJMSDJKAYCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product 442784 has been discontinued. Product PHR1417 may be an acceptable replacement.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

424.4 °F - closed cup

Flash Point(C)

218 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Squalane is in the midplane of the lipid bilayer: implications for its function as a proton permeability barrier.
Hau? T, et al.
Biochimica et Biophysica Acta - Bioenergetics, 1556(2-3), 149-154 (2002)
Biological importance and applications of squalene and squalane.
Advances in Food and Nutrition Research, 65(2-3), 223-233 (2012)
Brian K Radak et al.
The journal of physical chemistry. A, 113(26), 7218-7226 (2009-03-28)
To better understand the reactivity of gases with liquid surfaces, experimentalists have recently probed the reactive scattering of atomic fluorine at the surface of liquid squalane (C(30)H(62)). In this paper we further this research by simulating this scattering process at
Alexander M Zolot et al.
The Journal of chemical physics, 129(19), 194705-194705 (2008-11-26)
Exothermic chemical reaction dynamics at the gas-liquid interface have been investigated by colliding a supersonic beam of F atoms [E(com)=0.7(3) kcalmol] with a continuously refreshed liquid hydrocarbon (squalane) surface under high vacuum conditions. Absolute HF(v,J) product densities are determined by
L Harivardhan Reddy et al.
Molecular pharmaceutics, 6(5), 1526-1535 (2009-07-29)
Gemcitabine (2',2'-difluorodeoxyribofuranosylcytosine) is an anticancer nucleoside analogue active against a wide variety of solid tumors. However, following intravenous administration, this drug is rapidly inactivated by enzymatic deamination and displays a short biological half-life necessitating the administration of high doses leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service