Skip to Content
Merck
All Photos(1)

Documents

8.14605

Sigma-Aldrich

Squalane

for synthesis

Synonym(s):

Hexamethyltetracosane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.22

Quality Level

Assay

≥92.0% (GC)

form

liquid

bp

350 °C/1013 hPa

mp

-38 °C

transition temp

flash point 218 °C

density

0.81 g/cm3 at 20 °C

General description

for synthesis

Application

Squalane for synthesis. CAS 111-01-3, molar mass 422.81 g/mol.

Storage and Stability

Store below +30°C.

Analysis Note

Assay (GC, area%):≥ 92.0 % (a/a)
Density (d 20 °C/ 4 °C):0.808 - 0.814
Identity (IR):passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

424.4 °F - closed cup

Flash Point(C)

218 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service