Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1955

Sigma-Aldrich

Anti-MCM7 Antibody, clone 1C20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

DNA replication licensing factor MCM7;EC:3.6.4.12;CDC47 homolog;P1.1-MCM3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C20, recombinant monoclonal

description

1C20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 81.31 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

rat, human, monkey, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MCM7(4176)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects MCM7. It targets an epitope within 24 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the internal region of human MCM7.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MCM7 in HEK293 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MCM7 in COS-7, HeLa, NIH3T3, and C6 lysates.

Affinity Binding Assay: A representative lot of this antibody bound MCM7 peptide with a KD of 4.9 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MCM7 in HeLa cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MCM7 in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

DNA replication licensing factor MCM7 (UniProt: P33993; also known as EC:3.6.4.12, CDC47 homolog, P1.1-MCM3) is encoded by the MCM7 (also known as CDC47, MCM2) gene (Gene ID: 4176) in human. Unwinding of parental DNA in eukaryotic cells is performed by the 11-subunit Cdc45 MCM GINS assembly. MCM7 is a component of a highly conserved, multiprotein helicase complex that is required for DNA replication and G1/S transition. The six MCM proteins, MCM2-7 that belong to the AAA+ family of ATPases form a toroidal hetero-hexameric ring that translocates on single-strand DNA. In the absence of DNA substrate, MCM2-7 adopt an open spiral conformation. This complex serves as the replicative helicase essential for ′once per cell cycle′ DNA replication initiation and elongation in eukaryotic cells. MCM proteins bind with chromatin before the initiation of DNA replication and are then released from the chromatin as DNA replication proceeds. The active ATPase sites in the MCM2-7 ring are formed through the interaction surfaces of two neighboring subunits such that a critical structure of a conserved arginine finger motif (aa 513-516 in MCM7) is provided in trans relative to the ATP-binding site of the Walker A box of the adjacent subunit. The MCM domain of MCM7 is localized in amino acids 332-538. Three isoforms of MCM7 have been described that are produced by alternative splicing. Higher expression of MCM7 is reported in hepatocellular carcinoma (HCC) and is associated with worse overall survival. Knockdown of MCM7 is shown to inhibit cellular proliferation and HCC tumorigenicity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Rzechorzek, NJ., et al. (2020). Nucleic Acids Res. 48(12); 6980 6995; Qu, K., et al. (2017). Cell Death Dis. 8; 2603; Ishimi, Y. (1997). J. Biol. Chem. 272(39); 24508-24513).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.03 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service