Key Documents
Z381853
Ultramicro pipette tips
Microcapillary, flat tip, maximum volume 10 μL, non-sterile
Select a Size
1 PKG
R$1,019.00
R$1,019.00
Check Cart for Availability
Select a Size
Change View
1 PKG
R$1,019.00
About This Item
UNSPSC Code:
41121606
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
packaging:
pack of 200 ea (Bulk packed)
manufacturer/tradename:
Sorenson 1512
R$1,019.00
Check Cart for Availability
material
colorless
polypropylene
polypropylene
sterility
non-sterile
packaging
pack of 200 ea (Bulk packed)
manufacturer/tradename
Sorenson 1512
maximum volume
10 μL
volume
10 μL
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
For loading sample wells in 0.4 mm sequencing gels.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)