Skip to Content
Merck
All Photos(3)

Key Documents

WH0063974M6

Sigma-Aldrich

Monoclonal Anti-NEUROD6 antibody produced in mouse

clone 3B3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Atoh2, Anti-Math2, Anti-NEX1M, Anti-neurogenic differentiation 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,090.00

R$4,090.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$4,090.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,090.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEUROD6(63974)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
WH0003280M1AB9240WH0000474M1
biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

species reactivity

human

species reactivity

human

species reactivity

human, mouse

species reactivity

human

clone

3B3, monoclonal

clone

4D9, monoclonal

clone

polyclonal

clone

1B12, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

Gene Information

human ... NEUROD6(63974)

Gene Information

human ... HES1(3280)

Gene Information

mouse ... Neurod6(11922)

Gene Information

human ... ATOH1(474)

General description

NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM

Immunogen

NEUROD6 (NP_073565.2, 191 a.a. ~ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMH

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service