Skip to Content
Merck
All Photos(1)

Key Documents

Z186244

Sigma-Aldrich

Oxiclear disposable gas purifier

L × diam. 5 1/2 in. × 2 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$3,131.00

R$3,131.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
1 EA
R$3,131.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.51

R$3,131.00


Estimated to ship onApril 16, 2025


L × diam.

5 1/2 in. × 2 in.

General description

In-line purifier removes oxygen from inert carrier gases down to less than 50 ppb. Will purify 4 to 5 standard cylinders of 300 ft3. Delivers 100 cc/s at a pressure drop of 0.3 psi. Tested to 125 psi up to 100°C. Supplied with 1/4 in. compression fittings.

Legal Information

Oxiclear is a trademark of Labclear

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service