UC208
6-Hydroxywarfarin
Synonym(s):
3-(α-Acetonylbenzyl)-4,6-dihydrocoumarin, 4-6-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H16O5
CAS Number:
Molecular Weight:
324.33
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
Recommended Products
color
white
mp
119-120 °C
solubility
DMSO: soluble
storage temp.
2-8°C
SMILES string
CC(=O)CC(c1ccccc1)C2=C(O)c3cc(O)ccc3OC2=O
InChI
1S/C19H16O5/c1-11(20)9-14(12-5-3-2-4-6-12)17-18(22)15-10-13(21)7-8-16(15)24-19(17)23/h2-8,10,14,21-22H,9H2,1H3
InChI key
IQWPEJBUOJQPDE-UHFFFAOYSA-N
Biochem/physiol Actions
CYP1A2/2C9 metabolite of (R)-warfarin
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Preparation Note
6-Hydroxywarfarin is soluble in DMSO.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Development of a method for the analysis of warfarin and metabolites in plasma and urine.
J X de Vries et al.
American clinical laboratory, 14(7), 20-21 (1995-06-07)
Abdul Naveed Shaik et al.
Journal of pharmaceutical sciences, 105(6), 1976-1986 (2016-04-23)
The anticoagulant drug warfarin and the lipid-lowering statin drugs are commonly co-administered to patients with cardiovascular diseases. Clinically significant drug-drug interactions (DDIs) between these drugs have been recognized through case studies for many years, but the biochemical mechanisms causing these
A E Rettie et al.
Chemical research in toxicology, 5(1), 54-59 (1992-01-01)
Previous kinetic studies have identified a high-affinity (S)-warfarin 7-hydroxylase present in human liver microsomes which appears to be responsible for the termination of warfarin's biological activity. Inhibition of the formation of (S)-7-hydroxywarfarin, the inactive, major metabolite of racemic warfarin in
Steven Lane et al.
British journal of clinical pharmacology, 73(1), 66-76 (2011-06-23)
Warfarin is a drug with a narrow therapeutic index and large interindividual variability in daily dosing requirements. Patients commencing warfarin treatment are at risk of bleeding due to excessive anticoagulation caused by overdosing. The interindividual variability in dose requirements is
Stacie M Bratton et al.
The Journal of pharmacology and experimental therapeutics, 340(1), 46-55 (2011-10-06)
Coumadin (R-, S-warfarin) is a challenging drug to accurately dose, both initially and for maintenance, because of its narrow therapeutic range and wide interpatient variability and is typically administered as a racemic (Rac) mixture, which complicates the biotransformation pathways. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service