Key Documents
TMO436111
Nunc® 96-well immuno plate
polystyrene, non-sterile, without lid, white, case of 80 ea
Synonym(s):
assay microplate, surface treated microplate
Select a Size
80 EACH
R$4,714.00
R$4,714.00
Check Cart for Availability
Select a Size
Change View
80 EACH
R$4,714.00
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
R$4,714.00
Check Cart for Availability
material
polystyrene
white
sterility
non-sterile
packaging
case of 80 ea
manufacturer/tradename
Nunc 436111
General description
Thermo Scientific White 96-Well Immuno Plates are used to perform both quantitative and qualitative immunoassays, such as ELISA and binding assays. It is designed to be compatible with commonly used instruments. These plates ensure excellent performance, consistent results between different lots, and reproducibility across individual wells.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)