Skip to Content
Merck
All Photos(1)

Key Documents

M5785

Sigma-Aldrich

Nunc-Immuno MicroWell 96 well solid plates

Nunc-Immuno™ MicroWell™ 96 well solid plates

96 well Microwell MaxiSorp flat bottom plate, pinchbar design, with certificate

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, ELISA plates, cell culture plates, immunoassay plates, tissue culture plates

Sign Into View Organizational & Contract Pricing

Select a Size

60 EA
R$3,480.00

R$3,480.00


Available to ship onApril 07, 2025Details



Select a Size

Change View
60 EA
R$3,480.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear polystyrene wells
size:
96 wells

R$3,480.00


Available to ship onApril 07, 2025Details


material

clear polystyrene
flat bottom clear polystyrene wells

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 60 ea

manufacturer/tradename

Nunc 439454

plate L × W

128 mm × 86 mm

size

96 wells

surface area

2.7 cm2

total capacity

400 μL

well maximum volume

400 μm

working volume

350 μL

suitability

suitable for (quantitative/qualitative solid phase immuno assays (e.g. ELISA) and binding assays)

binding type

(MaxiSorp surface)
high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Nunc-Immuno 96 MicroWell solid plates are intended for quantitative and qualitative solid phase immuno assays (i.e. ELISA) and binding assays.

  • Raised rims on the wells
  • Certified reproducibility of binding
  • Alphanumeric orientation system
  • Fit standard equipment
  • Barcodes available on request

Surface Treaments:
  • PolySorp® surface treament, high affinity to molecules of a hydrophobic nature
  • MediSorp surface treament, surface chemistry between PolySorp and MaxiSorp, provides low background signal with samples containing serum
  • MaxiSorp surface treament, high affinity to molecules with mixed hydrophilic/hydrophobic domains
  • MultiSorp surface treament, high affinity to hydrophilic molecules
These plates are packaged in a sleeve of 60 plates and are not individually wrapped.

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
MediSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
MultiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc-Immuno is a trademark of Thermo Fisher Scientific or its subsidiaries
PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service