Skip to Content
Merck
All Photos(1)

Key Documents

TMO312185-0008

Nalgene® wide-mouth bottle

high-density polyethylene, capacity 250 mL, amber, case of 250 ea

Synonym(s):

light resistant container, opaque packaging bottle

Sign Into View Organizational & Contract Pricing

Select a Size

250 EACH
R$3,261.00

R$3,261.00


Check Cart for Availability


Select a Size

Change View
250 EACH
R$3,261.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

R$3,261.00


Check Cart for Availability

material

high-density polyethylene

packaging

case of 250 ea

manufacturer/tradename

Nalgene 312185-0008

capacity

250 mL

color

amber

Looking for similar products? Visit Product Comparison Guide

General description

Confidently ship and store these Thermo Scientific Nalgene Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service