B7285
Nalgene® bottles, style 2120
capacity 2,000 mL
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles
About This Item
Recommended Products
material
high-density polyethylene bottle
polypropylene cap
polypropylene screw closure
round bottle
Joint
Joints threaded neck (100)
description
Not autoclavable
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 2120-0005
bottle diam. × H
199 mm × 244 mm
capacity
2,000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Resistant to chemicals including acids, bases, and alcohols
- Can be stored at -100°C for deep freezing
- Suitable for long-term storage
- Non-leakproof and non-toxic
- Caps are liner-less that close the bottle without a liner that can wrinkle, leak, and contaminate the solution
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service