Skip to Content
Merck
All Photos(1)

Key Documents

T2452

Sigma-Aldrich

Talsaclidine

≥98% (HPLC), oil

Synonym(s):

(R)-3-(2-Propynyloxy)-1-Azabicyclo[2.2.2]octane, WAL 2014

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

C#CCO[C@H]1CN2CC[C@H]1CC2

InChI

1S/C10H15NO/c1-2-7-12-10-8-11-5-3-9(10)4-6-11/h1,9-10H,3-8H2/t10-/m0/s1

InChI key

XVFJONKUSLSKSW-JTQLQIEISA-N

Biochem/physiol Actions

Talsaclidine aids in promoting the nonamyloidogenic α-secretase pathway in model systems. It is more effective in stimulating muscarinic M 1 receptors than M 2 and M 3 receptors. In anesthetized guinea pigs, talsaclidine has no bronchospastic effects, but it produces contracture in isolated tracheal muscle. Talsaclidine is considered a potential candidate for cholinergic replacement therapy.
Talsaclidine is a M1 selective muscarinic receptor agonist.
Talsaclidine is a M1 selective muscarinic receptor agonist. Concentration-dependently, talsaclidine increases the release of APPs and dose-dependently. Talsaclidine decreases Aβ formation and decreases cerebrospinal fluid Aβ in AD patients.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Packaged under argon

Preparation Note

Talsaclidine is soluble in DMSO at a concentration that is greater than 5 mg/ml.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Wienrich et al.
Life sciences, 68(22-23), 2593-2600 (2001-06-08)
In functional pharmacological assays, talsaclidine has been described as a functionally preferential M1 agonist with full intrinsic activity, and less pronounced effects at M2- and M3 receptors. In accordance with this, cholinomimetic central activation measured in rabbits by EEG recordings
C Hock et al.
Annals of the New York Academy of Sciences, 920, 285-291 (2001-02-24)
Brain amyloid load in Alzheimer's disease (AD) is, at least in genetic forms, associated with overproduction of amyloid beta-peptides (A beta). Thus, lowering A beta production is a central therapeutic target in AD and may be achieved by modulating such
A Walland et al.
European journal of pharmacology, 330(2-3), 213-219 (1997-07-09)
The aim of the present investigation was to determine the reasons why the muscarinic receptor agonist talsaclidine (WAL 2014 FU, 1-azabicyclo[2.2.2] octane,3-(2-propynyloxy)-, (R)-,(E)-2-butenedioate) is devoid of bronchospastic effects in anaesthetized guinea pigs but causes contracture in isolated tracheal muscle from

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service