Skip to Content
Merck
All Photos(2)

Key Documents

857463P

Avanti

DGTS-d9

Avanti Research - A Croda Brand 857463P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-O-4′-[N,N,N-trimethyl(d9)]-homoserine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$901.00

R$901.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$901.00

About This Item

Empirical Formula (Hill Notation):
C42H72D9NO7
CAS Number:
Molecular Weight:
721.15
UNSPSC Code:
12352211
NACRES:
NA.25

R$901.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857463P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857463P

lipid type

deuterated lipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COCCC([N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([O-])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C42H81NO7/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-40(44)49-37-38(36-48-35-34-39(42(46)47)43(3,4)5)50-41(45)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38-39H,6-37H2,1-5H3/t38-,39?/m0/s1/i3D3,4D3,5D3

InChI key

JENDGLMWMJVGPR-AJAOUQJPSA-N

General description

1,2-dipalmitoyl-sn-glycero-3-O-4′-[N,N,N-trimethyl(d9)]-homoserine (DGTS) is highly found in Chlorophyceae. It is a structural isomer of diacylglycerylhydroxymethyl-N,N,N-trimethyl-βalanine (DGTA) It is also known as betaine lipid in microalgae.[1]
Diacylglyceryltrimethylhomo-Ser (DGTS) is an abundant lipid in the membranes of many algae, lower plants, and fungi. It commonly has an inverse concentration relationship with phosphatidylcholine, thus seemingly capable of replacing this phospholipid in these organisms.

Packaging

5 mL Amber Glass Screw Cap Vial (857463P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous structural identification of diacylglyceryl-N-trimethylhomoserine (DGTS) and diacylglycerylhydroxymethyl-N, N, N-trimethyl-$\beta$-alanine (DGTA) in microalgae using dual Li+/H+ adduct ion mode by ultra-performance liquid chromatography/quadrupole time-of-flight mass spectrometry
Li Y, et al.
Rapid Communications in Mass Spectrometry, 31(5), 457-468 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service