Skip to Content
Merck
All Photos(1)

Key Documents

SRP6427

Sigma-Aldrich

FGFR1/CD331 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

BFGFR, CD331, CEK, FGFBR, FGFR-1, FLT2, H2, HBGFR, KAL2, N-SAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 40.1 kDa
observed mol wt 60-65 kDa by SDS-PAGE (DTT-reduced. Arg 22 is the predicted N-terminal.)

packaging

pkg of 20 and 100 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGFR1(2260)

General description

Fibroblast growth factor receptor 1 (FGFR1) is also known as basic fibroblast growth factor receptor 1(BFGFR1), FMS-like tyrosine kinase, CD331, and is a receptor tyrosine kinase whose ligands are specific members of the fibroblast growth factor family. This protein is one of several fibroblast growth factor receptors, which are related proteins that are involved in important processes such as cell division, regulation of cell growth and maturation, formation of blood vessels, wound healing, and embryonic development. The FGFR1 protein spans the cell membrane, so that one end of the protein remains inside the cell and the other end projects from the outer surface of the cell. This positioning allows the FGFR1 protein to interact with specific growth factors outside the cell and to receive signals that help the cell respond to its environment. When growth factors attach to the FGFR1 protein, the receptor triggers a cascade of chemical reactions inside the cell that instruct the cell to undergo certain changes, such as maturing to take on specialized functions. The FGFR1 protein is thought to play an important role in the development of the nervous system. This protein may also help regulate the growth of long bones, such as the large bones in the arms and legs.
The FGFR1 (fibroblast growth factor receptor 1) gene is mapped to human chromosome 8p11.23. It codes for a cell-surface receptor tyrosine kinases and belongs to the FGFR family.

Biochem/physiol Actions

Fgfr1 (fibroblast growth factor receptor 1) protein prevents endothelial to mesenchymal transition by downregulating transforming growth factor β. Fgfr1 thus helps in maintaining normal vascularity, which is generally affected by diabetes condition. Upregulation of the FGFR1 gene indicates tumor progression and is observed in squamous cell carcinoma.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-affinity internalizing human scFv-fc antibody for targeting FGFR1-overexpressing lung cancer.
Sokolowska-Wedzina A, et al.
Molecular Cancer Research (2017)
FGFR1 is critical for the anti-endothelial mesenchymal transition effect of N-acetyl-seryl-aspartyl-lysyl-proline via induction of the MAP4K4 pathway.
Li J, et al.
Cell Death & Disease, 8(8), e2965-e2965 (2017)
A genome?wide linkage scan for cleft lip and cleft palate identifies a novel locus on 8p11?23.
Riley B M, et al.
American Journal of Medical Genetics. Part A, 143(8), 846-852 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service