Skip to Content
Merck
All Photos(1)

Key Documents

SRP3006

Sigma-Aldrich

AITRL human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Activation-induced TNFR member Ligand, TNFSF18

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$3,141.00

R$3,141.00


Estimated to ship onMay 20, 2025



Select a Size

Change View
20 μG
R$3,141.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$3,141.00


Estimated to ship onMay 20, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

14.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFSF18(8995)

General description

AITRL, a member of the TNF superfamily, is expressed in endothelial cells, and signals through the AITR receptor. AITRL regulates T-cell proliferation and survival, and effectuates the interaction between T lymphocytes and endothelial cells. The AITRL gene codes for a type II transmembrane protein comprised of 177 amino acids, including a 28 amino acid cytoplasmic region, a 21 amino acid transmembrane domain and a 128 amino acid extracellular domain. Recombinant human soluble AITRL is a 14.3 kDa protein, containing 126 amino acid residues corresponding to the extracellular domain of AITRL.

Biochem/physiol Actions

AITRL, a member of the TNF superfamily, is expressed in endothelial cells, and signals through the AITR receptor. Recombinant human soluble AITRL is a 14.3 kDa protein, containing 126 amino acid residues corresponding to the extracellular domain of AITRL.

Sequence

ETAKEPCMAK FGPLPSKWQM ASSEPPCVNK VSDWKLEILQ NGLYLIYGQV APNANYNDVA PFEVRLYKNK DMIQTLTNKS KIQNVGGTYE LHVGDTIDLI FNSEHQVLKN NTYWGIILLA NPQFIS

Physical form

Lyophilized from 10 mM Tris, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Gurney et al.
Current biology : CB, 9(4), 215-218 (1999-03-13)
The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate diverse biological functions, including cell proliferation, differentiation, and survival [1] [2] [3]. We have identified a new TNF-related ligand, designated human GITR ligand (hGITRL), and its human receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service