Skip to Content
Merck
All Photos(1)

Key Documents

SRP0690

Sigma-Aldrich

LKB1(STK11) Protein

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$3,827.00

R$3,827.00


Check Cart for Availability


Select a Size

Change View
50 μG
R$3,827.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

R$3,827.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~85 kDa

concentration

0.2 μg/μL

accession no.

NM_000455

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LKB1(6794)

Application

Useful for Western blot and in kinases assays as a downstream substrate for SRC or MEK1 kinases.

Biochem/physiol Actions

Recombinant full-length human LKB1(STK11) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. LKB1(STK11) is a member of the serine/threonine kinase family which regulates cell polarity and functions as a tumor suppressor. STK11 is a critical barrier to pulmonary tumorigenesis which controlling initiation, differentiation, and metastasis. STK11 plays a central role in restricting HSC entry into cell cycle and in broadly maintaining energy homeostasis in hematopoietic cells through a novel metabolic checkpoint (1). STK11 serves as an essential regulator of HSCs and hematopoiesis, and more generally, points to the critical importance of coupling energy metabolism and stem cell homeostasis (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service