Skip to Content
Merck
All Photos(1)

Key Documents

14-840-M

Sigma-Aldrich

AMPK (α1, β1, γ1) Protein, active, 10 µg

Active, recombinant, human AMPK (α1, β1, γ1) holoenzyme, comprised of full length N-terminal 6His-tagged AMPKα1, full length untagged AMPKβ1 & full length untagged AMPKγ1. For use in Kinase Assays.

Synonym(s):

AMPK kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

shelf life

6 mo.

mol wt

Mw 30 kDa (AMPKß1)
Mw 38 kDa (AMPKγ1)
Mw 64 kDa (AMPKα1)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

solubility

water: soluble

GenBank accession no.

NCBI accession no.

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... PRKAB1(5564)

General description

Research area: Cell Signaling

Product Source: Expressed in E. coli cells and activated in vivo.

Protein Target: AMPK (α1, β1, γ1)

Target Sub-Family: CAMK

Recombinant, human AMPK (α1, β1, γ1) holoenzyme, comprised of full- length N-terminal 6His-tagged AMPKα1, full length untagged AMPKβ1 and full length untagged AMPKγ1. The AMPK α catalytic subunit in mammals is encoded by α1 and α2, two β genes β1 and β2 and three γ subunit genes γ1, γ2, and γ3.[1]

Application

For use in Kinase Assays.
Research Category
Metabolism

Apoptosis & Cancer
Research Sub Category
Diabetes

Obesity

Metabolic Disorders

Biochem/physiol Actions

Under conditions of nutrient scarcity, AMPK functions as a metabolic checkpoint and inhibits cellular growth.[1] Upon activation, AMPK promotes glucose uptake and fatty acid oxidation thereby reducing energy-consuming processes like protein and lipid synthesis.[2] Furthermore, AMPK regulates autophagy and mitophagy.[3] It also facilitates catabolic pathways to generate more ATP as a response to decreased ATP production.[1]

Packaging

Also available in 250µg size as catalog # 14-840M - Call for pricing and availability.

Quality

Routinely evaluated by phosphorylation of substrate peptide

Physical form

Cobalt-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AMPK: guardian of metabolism and mitochondrial homeostasis
Herzig S and Shaw RJ
Molecular Cell Biology, 19, 121?135-121?135 (2018)
AMPK and the biochemistry of exercise: Implications for human health and disease
Richter EA and Ruderman NB
The Biochemical Journal, 418(2), 261?275-261?275 (2009)
The AMP-activated protein kinase (AMPK) signaling pathway coordinates cell growth, autophagy, & metabolism
Mihaylova MM and Shaw RJ
Nature Cell Biology, 13(9), 1016?1023-1016?1023 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service