Skip to Content
Merck
All Photos(2)

Key Documents

SRP0447

Sigma-Aldrich

OTUD6B active human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

CGI-77, DUBA5, OTU domain containing 6B

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$3,021.00

R$3,021.00


Estimated to ship onMay 22, 2025



Select a Size

Change View
50 μG
R$3,021.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

R$3,021.00


Estimated to ship onMay 22, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

39 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... OTUD6B(51633)

General description

Human deubiquitinating enzyme ovarian tumor domain- containing 6B, or OTUD6B (GenBank Accession No. NM_016023) amino acids 2-323 (end) with N-terminal His-Flag-tag, MW=39 kDa, expressed in E. coli cells.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu, Z. et al.
PLoS ONE, e14514-e14514 (2011)
Mathew E Sowa et al.
Cell, 138(2), 389-403 (2009-07-21)
Deubiquitinating enzymes (Dubs) function to remove covalently attached ubiquitin from proteins, thereby controlling substrate activity and/or abundance. For most Dubs, their functions, targets, and regulation are poorly understood. To systematically investigate Dub function, we initiated a global proteomic analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service