Skip to Content
Merck
All Photos(1)

Documents

SML3740

Sigma-Aldrich

PY-60

≥98% (HPLC)

Synonym(s):

5-Phenyl-N-[3-(2-thiazolyl)propyl]-3-isoxazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O2S
CAS Number:
Molecular Weight:
313.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

PY-60 is a potent and specific activator of the transcriptional coactivator Yes-associated protein 1 (YAP) signaling. It stimulates YAP-dependent expansion of epidermal keratinocytes in mouse. PY-60 binds directly to annexin A2 (ANXA2) causing its liberation and YAP activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service