Skip to Content
Merck
All Photos(1)

Key Documents

SML0849

Sigma-Aldrich

GKA50

≥98% (HPLC)

Synonym(s):

6-[[3-[(1S)-2-Methoxy-1-methylethoxy]-5-[(1S)-1-methyl-2-phenylethoxy]benzoyl]amino]-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$998.00
25 MG
R$5,307.00

R$998.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$998.00
25 MG
R$5,307.00

About This Item

Empirical Formula (Hill Notation):
C26H28N2O6
CAS Number:
Molecular Weight:
464.51
UNSPSC Code:
51111800
NACRES:
NA.77

R$998.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

N(c3ncc(cc3)C(=O)O)C(=O)c1cc(cc(c1)O[C@H](Cc2ccccc2)C)O[C@H](COC)C

InChI

1S/C26H28N2O6/c1-17(11-19-7-5-4-6-8-19)33-22-12-21(13-23(14-22)34-18(2)16-32-3)25(29)28-24-10-9-20(15-27-24)26(30)31/h4-10,12-15,17-18H,11,16H2,1-3H3,(H,30,31)(H,27,28,29)/t17-,18-/m0/s1

InChI key

OCBMECSFDVUYQN-ROUUACIJSA-N

Biochem/physiol Actions

GKA50 is a specific activator of glucokinase (EC50 = 0.03 μM). In mouse MIN6 insulin-secreting cells and rat pancreatic islet cells, GKA50 elicits a leftward shift in response to sub-maximal glucose concentrations as measured by insulin secretion and elevation of intracellular calcium levels.
GKA50 is a specific activator of glucokinase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service