Skip to Content
Merck
All Photos(1)

Key Documents

SML2921

Sigma-Aldrich

LYN-1604 hydrochloride

≥98% (HPLC)

Synonym(s):

1-(4-(2-(2,4-Dichlorophenyl)-2-(naphthalen-2-ylmethoxy)ethyl)piperazin-1-yl)-2-(diisobutylamino)ethanone hydrochloride, 2-[bis(2-Methylpropyl)amino]-1-[4-[2-(2,4-dichlorophenyl)-2-(2-naphthalenylmethoxy)ethyl]-1-piperazinyl]-ethanone hydrochloride, UA3-02 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$934.00
25 MG
R$2,302.00

R$934.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$934.00
25 MG
R$2,302.00

About This Item

Empirical Formula (Hill Notation):
C33H43Cl2N3O2 · xHCl
CAS Number:
Molecular Weight:
584.62 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

R$934.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

LYN-1604 is a potent ULK1 (UNC-51-like kinase 1; Autophagy-related protein 1 homolog) activator (EC50 = 18.94 nM; 1.96-fold activation at 100 nM) that induces ULK complex (ULK1-mATG13-FIP200-ATG101)-dependent death in the triple-negative breast cancer (TNBC) MDA-MB-231 cultures (IC50 = 1.66 μM) involving both autophagy and apoptosis pathway effectors (e.g. ATF3, RAD21, and caspase-3). Intragastric administration is shown to suppress MDA-MB-231 xenograft tumor growth in mice in vivo (EC50 ~50 mg/kg/day on day 14). Mutagenesis and in silico docking studies identify Lys50, Leu53, and Tyr89 as important ULK1 residues that mediate LYN-1604 target site interaction.
Potent ULK1 (UNC-51-like kinase 1; hTAG1) activator with in vitro and in vivo anti-cancer efficacy against triple-negative breast cancer (TNBC) MDA-MB-231.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Ouyang et al.
Autophagy, 13(4), 777-778 (2017-02-07)
ULK1 (unc-51 like autophagy activating kinase 1) is well known to be required to initiate the macroautophagy/autophagy process, and thus activation of ULK1-modulating autophagy/autophagy-associated cell death (ACD) may be a possible therapeutic strategy in triple negative breast cancer (TNBC). Here
Shisi Ma et al.
Experimental eye research, 184, 126-134 (2019-04-25)
Dry eye disease (DED), a multifactorial ocular surface disorder affecting millions of individuals worldwide, is characterized by inflammation and damage to the ocular surface. It is unclear whether corneal autophagy participates in ocular surface inflammation observed in DED. To test
Lan Zhang et al.
Chemical science, 8(4), 2687-2701 (2017-05-30)
UNC-51-like kinase 1 (ULK1) is well-known to initiate autophagy, and the downregulation of ULK1 has been found in most breast cancer tissues. Thus, the activation of ULK1-modulated autophagy could be a promising strategy for breast cancer therapy. In this study
Bo Wang et al.
Molecular cell, 74(4), 742-757 (2019-04-14)
Disturbances in autophagy and stress granule dynamics have been implicated as potential mechanisms underlying inclusion body myopathy (IBM) and related disorders. Yet the roles of core autophagy proteins in IBM and stress granule dynamics remain poorly characterized. Here, we demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service