Skip to Content
Merck
All Photos(1)

Key Documents

SML1684

Sigma-Aldrich

GSK2850163

≥98% (HPLC)

Synonym(s):

(R)-2-(3,4-Dichlorobenzyl)-N-(4-methylbenzyl)-2,7-diazaspiro[4.5]decane-7-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$766.00
25 MG
R$3,075.00

R$766.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$766.00
25 MG
R$3,075.00

About This Item

Empirical Formula (Hill Notation):
C24H29Cl2N3O
CAS Number:
Molecular Weight:
446.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$766.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

SMILES string

ClC1=CC(CN(C2)CC[C@]32CN(C(NCC4=CC=C(C)C=C4)=O)CCC3)=CC=C1Cl

InChI

1S/C24H29Cl2N3O/c1-18-3-5-19(6-4-18)14-27-23(30)29-11-2-9-24(17-29)10-12-28(16-24)15-20-7-8-21(25)22(26)13-20/h3-8,13H,2,9-12,14-17H2,1H3,(H,27,30)/t24-/m1/s1

InChI key

YFDASBFQKMHSSJ-XMMPIXPASA-N

Biochem/physiol Actions

GSK2850163 is a highly selective inhibitor of inositol requiring enzyme-1 alpha (IRE1α) with dual activity: it inhibits IRE1α kinase activity with an IC50 value of 20 nM and RNase activity with an IC50 value of 200 nM. IRE1α is a dual serine/threonine-protein kinase/endoribonuclease with two enzymatic domains: a trans-autophosphorylation domain and and endoribonuclease (RNase) domain. When activated, usually in response to endoplasmic reticulum (ER) stress such as the unfolded protein response (UPR), IRE1α oligomerizes and activates mRNA splicing of adaptive XBP1 transcription factor, which upregulates ER chaperones and ER associated degradation (ERAD) genes that promote degradation of ER unfolded proteins and facilitate recovery from ER stress. However, if ER stress is too high or chronic, IRE1α is hyperactivated, increasing many ER mRNAs, resulting in apoptosis. GSK2850163 interacts with IRE1α catalytic residues Lys599 and Glu612, displacing the kinase activation loop to the inactive DFG-out conformation. The rearrangement of the kinase domain-dimer interface also results in the RNAse domains rotating away from each other, inhibiting the RNase activity.
IRE1α-selective inhibitor of XBP1 splicing
IRE1α-selective inhibitor of XBP1 splicing.
Homoharringtonine is a cephalotaxine ester that is also known as (HHT; 4-methyl (2R)-2-hydroxy-2-(4-hydroxy-4-methylpentyl)butanedioate). In gefitinib-resistant lung cancer cells, homoharringtonine promotes apoptosis and prevents signal transducer and activator of transcription (STAT3) through IL-6 (interleukin-6) /JAK1 (janus kinase 1)/STAT3 signal pathway.[1] It plays an important role in the treatment of malaria.[2]

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemistry and Biology, 439-439 (1998)
Wei Cao et al.
Scientific reports, 5, 8477-8477 (2015-07-15)
Tyrosine kinase inhibitors (TKIs) are mostly used in non-small cell lung cancer (NSCLC) treatment. Unfortunately, treatment with Gefitinib for a period of time will result in drug resistance and cause treatment failure in clinic. Therefore, exploring novel compounds to overcome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service