Skip to Content
Merck
All Photos(1)

Key Documents

SML2490

Sigma-Aldrich

APC366 trifluoroacetate

≥97% (HPLC)

Synonym(s):

APC 366 trifluoroacetate, N-(1-Hydroxy-2-naphthoyl)-L-arginyl-L-prolinamide, N2-[(1-Hydroxy-2-naphthalenyl)carbonyl]-L-arginyl-L-prolinamide trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$711.00
25 MG
R$1,790.00

R$711.00


Estimated to ship onApril 18, 2025



Select a Size

Change View
5 MG
R$711.00
25 MG
R$1,790.00

About This Item

Empirical Formula (Hill Notation):
C22H28N6O4 · xC2HF3O2
CAS Number:
Molecular Weight:
440.50 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

R$711.00


Estimated to ship onApril 18, 2025


Assay

≥97% (HPLC)

form

lyophilized powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

FC(F)(C(O)=O)F.NC(NCCC[C@@H](C(N1CCC[C@H]1C(N)=O)=O)NC(C2=C(O)C3=CC=CC=C3C=C2)=O)=N

InChI

1S/C22H28N6O4/c23-19(30)17-8-4-12-28(17)21(32)16(7-3-11-26-22(24)25)27-20(31)15-10-9-13-5-1-2-6-14(13)18(15)29/h1-2,5-6,9-10,16-17,29H,3-4,7-8,11-12H2,(H2,23,30)(H,27,31)(H4,24,25,26)/t16-,17-/m0/s1

InChI key

SKYWIMYOGAWOMB-IRXDYDNUSA-N

Biochem/physiol Actions

APC366 is a selective inhibitor of mast cell tryptase, which is involved with allergenic response. It thereby also inhibits protease-activated receptor PAR2, a G-protein-coupled receptor that is activated by mast cell tryptase. Tryptase has been proposed to be involved in fibrosis, joint inflammation and also in promoting breast cancer angiogenesis, all of which APC366 inhibited.
Mast cell tryptase inhibitor

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neng Qian et al.
Oncology letters, 16(2), 1513-1520 (2018-07-17)
Mast cells have been demonstrated to accumulate around and within solid tumors of numerous types, and express a number of pro-angiogenic compounds, including tryptase. They may serve an early role in angiogenesis within developing tumors. In the present study, the
Issan Yee San Tam et al.
International archives of allergy and immunology, 177(3), 199-206 (2018-07-19)
Mast cells are key immune effector cells which release chemokines, proteases, and other inflammatory mediators upon activation by immunological stimuli. The aim of this study was to investigate the effects of co-releasing proteases on the kinetics of release of the
Alexandre Denadai-Souza et al.
Arthritis research & therapy, 19(1), 124-124 (2017-06-08)
Increasing evidences indicate that an unbalance between tryptases and their endogenous inhibitors, leading to an increased proteolytic activity, is implicated in the pathophysiology of rheumatoid arthritis. The aim of the present study was to evaluate the impact of tryptase inhibition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service