Skip to Content
Merck
All Photos(1)

Key Documents

SML2155

Sigma-Aldrich

TAK-901

≥98% (HPLC)

Synonym(s):

5-[3-(Ethylsulfonyl)phenyl]-3,8-dimethyl-N-(1-methyl-4-piperidinyl)-9H-pyrido[2,3-b]indole-7-carboxamide, TAK 901

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$733.00
25 MG
R$2,382.00

R$733.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$733.00
25 MG
R$2,382.00

About This Item

Empirical Formula (Hill Notation):
C28H32N4O3S
CAS Number:
Molecular Weight:
504.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

R$733.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(CC)c1cc(ccc1)c2c3c([nH]c5ncc(cc53)C)c(c(c2)C(=O)NC4CCN(CC4)C)C

InChI

1S/C28H32N4O3S/c1-5-36(34,35)21-8-6-7-19(14-21)23-15-22(28(33)30-20-9-11-32(4)12-10-20)18(3)26-25(23)24-13-17(2)16-29-27(24)31-26/h6-8,13-16,20H,5,9-12H2,1-4H3,(H,29,31)(H,30,33)

InChI key

WKDACQVEJIVHMZ-UHFFFAOYSA-N

Biochem/physiol Actions

TAK-901 is a potent and tight-binding Aurora B kinase inhibitor. TAK-901 targets multiple kinases in vitro, but appears to mainly target Aurora B in cells and in vivo. TAK-901 suppresses cellular histone H3 phosphorylation and induced polyploidy.
potent and tight-binding Aurora B kinase inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saomi Murai et al.
Anticancer research, 37(2), 437-444 (2017-02-10)
Aurora B kinase plays an essential role in chromosome segregation and cytokinesis, and is dysregulated in many cancer types, making it an attractive therapeutic target. TAK-901 is a potent aurora B inhibitor that showed efficacy in both in vitro and
Frances V Hundley et al.
Molecular cell, 81(6), 1319-1336 (2021-02-05)
The human ubiquitin proteasome system, composed of over 700 ubiquitin ligases (E3s) and deubiquitinases (DUBs), has been difficult to characterize systematically and phenotypically. We performed chemical-genetic CRISPR-Cas9 screens to identify E3s/DUBs whose loss renders cells sensitive or resistant to 41

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service