Skip to Content
Merck
All Photos(1)

Key Documents

SML2672

Sigma-Aldrich

Tandutinib hydrochloride

≥98% (HPLC)

Synonym(s):

4-[6-Methoxy-7-[3-(1-piperidinyl)propoxy]-4-quinazolinyl]-N-[4-(1-methylethoxy)phenyl]-1-piperazinecarboxamide hydrochloride, CT 53518 hydrochloride, CT-53518 hydrochloride, CT53518 hydrochloride, MLN 518 hydrochloride, MLN-518 hydrochloride, MLN518 hydrochloride, N-(4-Isopropoxyphenyl)-4-(6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-yl)piperazine-1-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$473.00
100 MG
R$766.00

R$473.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
R$473.00
100 MG
R$766.00

About This Item

Empirical Formula (Hill Notation):
C31H42N6O4 · xHCl
CAS Number:
Molecular Weight:
562.70 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

R$473.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N5(CCCCC5)CCCOc1cc2ncnc(c2cc1OC)N3CCN(CC3)C(=O)Nc4ccc(cc4)OC(C)C

InChI

1S/C31H42N6O4/c1-23(2)41-25-10-8-24(9-11-25)34-31(38)37-17-15-36(16-18-37)30-26-20-28(39-3)29(21-27(26)32-22-33-30)40-19-7-14-35-12-5-4-6-13-35/h8-11,20-23H,4-7,12-19H2,1-3H3,(H,34,38)

InChI key

UXXQOJXBIDBUAC-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent βPDGFR, c-Kit, Flt3 (Flk2, Stk1) receptor tyrosine kinase inhibitor with anti-leukemia efficacy in vitro and in vivo.
Tandutinib (CT53518, MLN518) is a potent receptor tyrosine kinase inhibitor against PDGFRβ, c-Kit, Flt3 (IC50 = 0.20, 0.17, 0.22, 3.43 μM, respectively, against 8 nM PDGF-BB induced autophosphorylation of βPDGFR, βPDGFR/c-Kit, βPDGFR/Flt3, βPDGFR/CSF-1R chimera expressed in CHO cells; IC50 =30 μM against KDR, EFGR, FGFR, InsR, Abl, Src, PKA, PKC, ERK2, MEK1, MKK4/6, p38). Tandutinib prolongs the survival in murine CMML (Baf3 with Tel/βPDGFR chimera) and Flt-3/ITD AML leukemia models (60 mg/kg bid p.o.) with good oral bioavailability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nimitha R Mathew et al.
Nature medicine, 24(3), 282-291 (2018-02-13)
Individuals with acute myeloid leukemia (AML) harboring an internal tandem duplication (ITD) in the gene encoding Fms-related tyrosine kinase 3 (FLT3) who relapse after allogeneic hematopoietic cell transplantation (allo-HCT) have a 1-year survival rate below 20%. We observed that sorafenib
Sivapriya Ponnurangam et al.
Molecular cancer therapeutics, 12(5), 598-609 (2013-02-22)
The c-Kit receptor can activate distinct signaling pathways including phosphoinositide 3-kinase (PI3K)/Akt and mTOR. Aberrant c-Kit activation protects cells from apoptosis and enhances invasion of colon carcinoma cells. Tandutinib is a novel quinazoline-based inhibitor of the type III receptor tyrosine
Louise M Kelly et al.
Cancer cell, 1(5), 421-432 (2002-07-19)
Up to 30% of acute myelogenous leukemia (AML) patients harbor an activating internal tandem duplication (ITD) within the juxtamembrane domain of the FLT3 receptor, suggesting that it may be a target for kinase inhibitor therapy. For this purpose we have
Yousef Salama et al.
Biochemical and biophysical research communications, 490(2), 209-216 (2017-06-12)
Thymic regeneration is a crucial function that allows for the generation of mature T cells after myelosuppression like irradiation. However molecular drivers involved in this process remain undefined. Here, we report that the angiogenic factor, epidermal growth factor-like domain 7
Wen Deng et al.
Oncology reports, 29(6), 2479-2485 (2013-03-26)
It is well established that ATP-binding cassette (ABC) transporter-mediated multidrug resistance (MDR) is one of the major mechanisms that causes resistance to antineoplastic drugs in cancer cells. ABC transporters can significantly decrease the intracellular concentration of antineoplastic drugs by increasing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service