Skip to Content
Merck
All Photos(1)

Key Documents

SML1800

Sigma-Aldrich

JK-2

≥90% (NMR)

Synonym(s):

Lithium 2-(phenylphosphonothioylamino)-3-phenylpropanoate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$694.00
25 MG
R$2,791.00

R$694.00


Estimated to ship onApril 12, 2025



Select a Size

Change View
5 MG
R$694.00
25 MG
R$2,791.00

About This Item

Empirical Formula (Hill Notation):
C15H14NO3PS · 2Li
Molecular Weight:
333.20
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$694.00


Estimated to ship onApril 12, 2025


Quality Level

Assay

≥90% (NMR)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=P(C1=CC=CC=C1)([O-])NC(C([O-])=O)CC2=CC=CC=C2.[Li+].[Li+]

InChI

1S/C15H16NO3PS.2Li/c17-15(18)14(11-12-7-3-1-4-8-12)16-20(19,21)13-9-5-2-6-10-13;;/h1-10,14H,11H2,(H,17,18)(H2,16,19,21);;/q;2*+1/p-2

InChI key

JYJXLPYKDMREAV-UHFFFAOYSA-L

Biochem/physiol Actions

JK-2 is a phosphorothioate-based hydrogen sulfide (H2S) donor that shows greatly accelerated H2S release upon protonation-induced intramolecular cyclization at acidic pH ([H2S]/time/pH = 94 μM/4.5 min/pH 5.0, 84 μM/4.5 min/pH 6.0, 36 μM/90 min/pH 7.4, 21 μM/90 min/pH 8.0 with JK-2; [JK-2] = 100 μM at time 0) and is superior to GYY4137 in H2S generation both in buffer (<5 μM H2S/90 min/pH 5.0-8.0 with GYY4137) and in cells without cytotoxicity up to 400 μM in H9c2 and HeLa cultures. JK-2 protects H9c2 rat myoblasts from anoxia/reoxygenation (A/R; 1 hr 2% O2, then 6 hr reoxygenation with 400 μM H2O2) treatment (100% vs. 60% viability with or without 25 μM JK-2) and is efficacious in reducing infarct size per area-at-risk (INF/AAR) upon myocardial ischemia-reperfusion (MI/R) injury induction in mice in vivo (55% and 56% INF/AAR redcution with 50 μg/kg and 100 μg/kg, respectively, via intracardiac injection).
JK-2 is a phosphorothioate-based hydrogen sulfide (H2S) donor..

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service