Skip to Content
Merck
All Photos(1)

Key Documents

SML1418

Sigma-Aldrich

AC187 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

AC 187, AC-187, Ac-VLGKLSQELHKLQTYPRTNTGSNTY-NH2, Ac-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Asn-Thr-Tyr-NH2, N-Acetyl-30-L-asparagine-32-L-tyrosinamide-8-32-calcitonin (salmon)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C127H205N37O40 · xC2HF3O2
CAS Number:
Molecular Weight:
2890.21 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)C(C)C)CC(C)C)CCCCN)CC(C)C)CO)CCC(=O)N)CCC(=O)O)CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(=O)N)C(=O

InChI

1S/C127H205N37O40/c1-59(2)44-81(156-122(200)99(63(9)10)142-68(15)171)105(183)139-54-96(178)143-74(22-16-18-40-128)106(184)151-84(47-62(7)8)114(192)159-90(57-166)119(197)148-77(34-37-92(130)174)108(186)146-79(36-39-98(180)181)109(187)153-83(46-61(5)6)113(191)154-85(50-71-53-137-58-141-71)115(193)145-75(23-17-19-41-129)107(185)152-82(45-60(3)4)112(190)147-78(35-38-93(131)175)111(189)162-103(67(14)170)125(203)158-88(49-70-28-32-73(173)33-29-70)126(204)164-43-21-25-91(164)120(198)149-76(24-20-42-138-127(135)136)110(188)161-102(66(13)169)124(202)157-87(52-95(133)177)117(195)160-100(64(11)167)121(199)140-55-97(179)144-89(56-165)118(196)155-86(51-94(132)176)116(194)163-101(65(12)168)123(201)150-80(104(134)182)48-69-26-30-72(172)31-27-69/h26-33,53,58-67,74-91,99-103,165-170,172-173H,16-25,34-52,54-57,128-129H2,1-15H3,(H2,130,174)(H2,131,175)(H2,132,176)(H2,133,177)(H2,134,182)(H,137,141)(H,139,183)(H,140,199)(H,142,171)(H,143,178)(H,144,179)(H,145,193)(H,146,186)(H,147,190)(H,148,197)(H,149,198)(H,150,201)(H,151,184)(H,152,185)(H,153,187)(H,154,191)(H,155,196)(H,156,200)(H,157,202)(H,158,203)(H,159,192)(H,160,195)(H,161,188)(H,162,189)(H,163,194)(H,180,181)(H4,135,136,138)/t64-,65-,66-,67-,74+,75+,76+,77+,78+,79+,80+,81+,82+,83+,84+,85+,86+,87+,88+,89+,90+,91+,99+,100+,101+,102+,103+/m1/s1

InChI key

ZLFXHYNEZYAYPG-AABHONRUSA-N

Biochem/physiol Actions

AC187 is an orally active, potent and selective amylin receptor antagonist that increases glucagon secretion, alters plasma glucose levels and increases food intake in vivo. AC187 completely blocks internalization of amylin monomers in pancreatic cells. AC187 blocks β-amyloid neurotoxicity in rats.
AC187 is an orally active, potent and selective amylin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service