Skip to Content
Merck
All Photos(1)

Key Documents

SML1692

Sigma-Aldrich

AS8351

≥98% (HPLC)

Synonym(s):

2-Hydroxy-1-naphthylaldehyde isonicotinoylhydrazone, 311 Iron Chelator, AS-8351, NSC-51355

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$651.00

R$651.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$651.00

About This Item

Empirical Formula (Hill Notation):
C17H13N3O2
CAS Number:
Molecular Weight:
291.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$651.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1=CC=C(C=CC=C2)C2=C1/C=N/NC(C3=CC=NC=C3)=O

InChI

1S/C17H13N3O2/c21-16-6-5-12-3-1-2-4-14(12)15(16)11-19-20-17(22)13-7-9-18-10-8-13/h1-11,21H,(H,20,22)/b19-11+

InChI key

WQSHHAVECQJKLX-YBFXNURJSA-N

Biochem/physiol Actions

AS8351 is an iron chelator that has been found to inhibit of histone demethylase KDM5B (JARID1B), a histone H3K4 demethylase involved in regulation of cell proliferation and stem cell self-renewal and differentiation. AS8351 is thought to inhibit KDM5B by competing with a-ketoglutarate for chelating iron that KDM5B requires as a co-factor.
AS8351 is an iron chelator that has been found to inhibit of histone demethylase KDM5B (JARID1B).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service