Skip to Content
Merck
All Photos(1)

Documents

SMB00412

Sigma-Aldrich

2-Azidoethyl LacNAc

≥90% (HPLC)

Synonym(s):

β-D-LacNAc-Et-N3, 2-Azidoethyl β-D-Gal (1→4) β-D-GlcNAc, 2-Azidoethyl β-D-LacNAc, 2-Azidoethyl N-Acetyl-D-lactosamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N4O11
Molecular Weight:
452.41
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.79

Quality Level

Assay

≥90% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](NC(C)=O)[C@H](OCCN=[N+]=[N-])O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C16H28N4O11/c1-6(23)19-9-11(25)14(8(5-22)30-15(9)28-3-2-18-20-17)31-16-13(27)12(26)10(24)7(4-21)29-16/h7-16,21-22,24-27H,2-5H2,1H3,(H,19,23)/t7?,8?,9?,10-,11+,12-,13?,14+,15+,16-/m0/s1

InChI key

GBIYEXBPCNHQHL-WHWQPXQJSA-N

Related Categories

Biochem/physiol Actions

2-Azidoethyl LacNAc is an alkylated glycoside. Alkylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senlian Hong et al.
Cell chemical biology, 27(9), 1140-1150 (2020-07-11)
Wnt/β-catenin signaling regulates critical, context-dependent transcription in numerous physiological events. Among the well-documented mechanisms affecting Wnt/β-catenin activity, modification of N-glycans by L-fucose is the newest and the least understood. Using a combination of Chinese hamster ovary cell mutants with different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service