Skip to Content
Merck
All Photos(1)

Key Documents

SCP0170

Sigma-Aldrich

Granulocyte Elastase Substrate human

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$926.00

R$926.00


Estimated to ship onApril 18, 2025



Select a Size

Change View
10 MG
R$926.00

About This Item

Empirical Formula (Hill Notation):
C21H27N5O6
Molecular Weight:
445.47
UNSPSC Code:
12352204
NACRES:
NA.32

R$926.00


Estimated to ship onApril 18, 2025


Product Name

Granulocyte Elastase Substrate human,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Related Categories

Amino Acid Sequence

Glp-Pro-Val-pNA

General description

Elastase, a glycoprotein, is a member of the serine proteases family.[1] It is synthesized by human polymorphonuclear (PMN) leukocytes or by the pancreas.[2] Human granulocyte elastase is liberated in large amounts from granulocytes in chronic inflammatory joint disorders.[3]

Biochem/physiol Actions

Elastase is a catalytic enzyme that catalyzes the cleavage of carboxyl groups on tiny hydrophobic amino acids including glycine, alanine, and valine. Its main function is to break down elastin.[4] Free elastase in some purulent synovial fluids plays a key role in articular cartilage breakdown. Increased elastase levels in the plasma have been seen in patients with evident rheumatoid arthritis.[3]
Elastase(s) can be assayed and differentiated using chromogenic and fluorogenic Pro-Val peptide substrates such as Glp-Pro-Val-pNA and MeOSuc-Ala-Ala-Pro-Val-4-MNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of exocrine pancreatic function
Cotton SW
Contemporary Practice in Clinical Chemistry, 573-585 (2020)
Use of Extracts from Squirting Cucumber (Ecballium elaterium) Seeds in Health
Attard E
Nuts and Seeds in Health and Disease Prevention, 1079-1086 (2011)
K Kleesiek et al.
Rheumatology international, 6(4), 161-169 (1986-01-01)
Human granulocyte elastase (EC 3.4.21.37) is released from granulocytes in large amounts in chronic inflammatory joint diseases and is therefore of special pathogenic and diagnostic importance. In order to examine the diagnostic significance of this enzyme as a clinico-chemical parameter
Effects of Nigella (Nigella sativa L.) Seed Extract on Human Neutrophil Elastase Activity
Kacem R
Nuts and Seeds in Health and Disease Prevention, 823-829 (2011)
Pier Carlo Braga et al.
Pharmacology, 77(3), 150-154 (2006-07-11)
Erdosteine is a mucolytic drug whose metabolization gives rise to an active metabolite with an SH group (Met I), which reduces neutrophil release of reactive oxygen species and the peroxynitrite generated by the reaction of superoxide anion with nitric oxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service