Skip to Content
Merck
All Photos(1)

Key Documents

SCP0006

Sigma-Aldrich

Ac-Arg-Ala-Arg-AMC

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,203.00

R$2,203.00


Check Cart for Availability


Select a Size

Change View
5 MG
R$2,203.00

About This Item

Empirical Formula (Hill Notation):
C27H40N10O6
Molecular Weight:
600.67
UNSPSC Code:
12352202
NACRES:
NA.32

R$2,203.00


Check Cart for Availability

Product Name

Ac-Arg-Ala-Arg-AMC,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥62%

storage condition

protect from light

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service