Skip to Content
Merck
All Photos(1)

Key Documents

SCP0029

Sigma-Aldrich

[Gly22]-Amyloid β 1-40 Arctic human

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
R$2,120.00
1 MG
R$3,077.00

R$2,120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
R$2,120.00
1 MG
R$3,077.00

About This Item

Empirical Formula (Hill Notation):
C191H291N53O56S1
Molecular Weight:
4257.74
UNSPSC Code:
12352209
NACRES:
NA.77

R$2,120.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gly-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

General description

β-Amyloid peptides are the major constituents of senile plaques and neurofibrillary tangles that occur in the hippocampus, neocortex, and amygdala of patients with Alzheimer′s disease. Aβ (1-40) whether soluble or insoluble differentiates AD vs high pathology controls. The Arctic mutation E22G causes early onset of Alzheimer′s compared to wild type and promotes protofibril formation and neurotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Itkin et al.
PloS one, 6(3), e18250-e18250 (2011-04-06)
Amyloid β-peptide (Aβ) is directly linked to Alzheimer's disease (AD). In its monomeric form, Aβ aggregates to produce fibrils and a range of oligomers, the latter being the most neurotoxic. Dysregulation of Ca(2+) homeostasis in aging brains and in neurodegenerative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service