Skip to Content
Merck
All Photos(1)

Key Documents

A2326

Sigma-Aldrich

Amyloid β-Protein Fragment 40-1

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
R$1,815.00

R$1,815.00


Estimated to ship onJune 01, 2025



Select a Size

Change View
0.1 MG
R$1,815.00

About This Item

Empirical Formula (Hill Notation):
C194H295N53O58S
CAS Number:
Molecular Weight:
4329.80
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

R$1,815.00


Estimated to ship onJune 01, 2025


Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Amino Acid Sequence

Val-Val-Gly-Gly-Val-Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly-Val-Asp-Glu-Ala-Phe-Phe-Val-Leu-Lys-Gln-His-His-Val-Glu-Tyr-Gly-Ser-Asp-His-Arg-Phe-Glu-Ala-Asp

Biochem/physiol Actions

Inactive control against Αβ [1-40].

Other Notes

The N−C inverted sequence of Αβ [1-40].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W Kowall et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(16), 7247-7251 (1991-08-15)
Deposition of the beta-amyloid protein in senile plaques is a pathologic hallmark of Alzheimer disease (AD). Focal deposition of beta amyloid in the adult rat cerebral cortex caused profound neurodegenerative changes, including neuronal loss and degenerating neurons and neurites. Chronic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service