Skip to Content
Merck
All Photos(3)

Documents

SAB4502623

Sigma-Aldrich

Anti-BCL-XL antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Apoptosis regulator Bcl-X, BCL2L, BCL2L1, BCLX, Bcl-2-like 1 protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)

General description

Anti-BCL-XL Antibody detects endogenous levels of total BCL-XL protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human BCL-XL.

Immunogen Range: 13-62

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Stalin et al.
Oncogene, 35(42), 5489-5500 (2016-10-21)
CD146 (MUC-18, MCAM) expression on cancer cells correlates with cancer progression and a bad prognosis in several tumors, including melanoma and pancreatic tumors. Deciphering the mechanism mediating the CD146 role in cancer is essential for generating new therapeutic strategies. We
Jikui Xiong et al.
Oncology letters, 15(4), 4767-4774 (2018-03-20)
Malignant melanoma is characterized by rapid deterioration, early metastasis and high mortality. Cdk5 regulatory subunit-associated protein 1 (CDK5RAP1), which catalyzes 2-methylthio (ms2) modification of mitochondrial transfer RNAs, has been reported to induce cancer cell apoptosis, by a phospho-c-Jun N-terminal kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service