Skip to Content
Merck
All Photos(3)

Key Documents

SAB2100693

Sigma-Aldrich

Anti-EPOR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Erythropoietin receptor, Anti-MGC138358

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,042.00

R$3,042.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,042.00

About This Item

MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.41

R$3,042.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

human, mouse, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

SMILES string

O(C(=O)CCCCCCCCC\C=C/CCCCCCCC)C(=O)CCCCCCCCC\C=C/CCCCCCCC

InChI

1S/C40H74O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(41)43-40(42)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-38H2,1-2H3/b19-17-,20-18-

InChI key

QLXNJYBNAORLFC-CLFAGFIQSA-N

Gene Information

human ... EPOR(2057)

Immunogen

Synthetic peptide directed towards the N terminal region of human EPOR

Application

Anti-EPOR antibody produced in rabbit has been used in immunoblot analysis and magnetic cell sorting.[1]

Biochem/physiol Actions

The erythropoietin receptor is a member of the cytokine receptor family. Upon erythropoietin binding, the erythropoietin receptor activates Jak2 tyrosine kinase which activates different intracellular pathways including: Ras/MAP kinase, phosphatidylinositol 3-kinase and STAT transcription factors. The stimulated erythropoietin receptor appears to have a role in erythroid cell survival. Defects in the erythropoietin receptor may produce erythroleukemia and familial erythrocytosis.The erythropoietin receptor is a member of the cytokine receptor family. Upon erythropoietin binding, the erythropoietin receptor activates Jak2 tyrosine kinase which activates different intracellular pathways including: Ras/MAP kinase, phosphatidylinositol 3-kinase and STAT transcription factors. The stimulated erythropoietin receptor appears to have a role in erythroid cell survival. Defects in the erythropoietin receptor may produce erythroleukemia and familial erythrocytosis. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: DHLGASLWPQVGSLCLLLAGAAWAPPPNLPDPKFESKAALLAARGPEELL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Patapia Zafiriou et al.
Stem cells (Dayton, Ohio), 32(9), 2480-2491 (2014-05-09)
The role of erythropoietin (Epo) in myocardial repair after infarction remains inconclusive. We observed high Epo receptor (EPOR) expression in cardiac progenitor cells (CPCs). Therefore, we aimed to characterize these cells and elucidate their contribution to myocardial regeneration on Epo

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service