Skip to Content
Merck
All Photos(17)

Documents

ZRB1366

Sigma-Aldrich

Anti-Erythropoietin Antibody, clone 6E5, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EPO, Epoetin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6E5, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.31 kDa
observed mol wt ~20 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EPO(2056)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 6E5 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Erythropoietin.

Immunogen

His-tagged full-length mature recombinant human Erythropoietin (without the signal peptide).

Application

Anti-Erythropoietin, clone 6E5 ZooMAb, Cat. No. ZRB1366, a recombinant Rabbit Monoclonal Antibody that targets Erythropoietin and is tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Erythropoietin in HUVEC and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Erythropoietin in human kidney and rat kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Erythropoietin (UniProt: P01588; also known as Epoetin, EPO) is encoded the EPO gene (Gene ID: 2056) in human. EPO is an acidic glycoprotein that is synthesized with a signal peptide (aa 1-27) that is subsequently cleaved off in the endoplasmic reticulum and the mature protein undergoes glycosylation in the Golgi prior to its release. EPO is mainly produced by peritubular fibroblasts in the renal cortex and to some extent by the liver in adult mammals. In fetal and neonatal mammals, it produced in the liver. EPO is the principle regulator of red blood cell production by stimulating the division and differentiation of bone marrow erythroid progenitor cells in response to anemia and hypoxia. It is induced by ischemic preconditioning by activation of (HIF)-1alpha. It is reported to contribute to angiogenesis in response to ischemia through upregulation of VEGF/VEGFR. It binds to EPO receptor that leads to receptor dimerization and JAK2 activation that results in activation of specific downstream effectors, including STAT1 and STAT3. Endogenous EPO consists of three N-linked polysaccharide groups and one O-linked group. Although its glycosylation slightly reduces its biological activity but is shown to be essential for ensuring its prolonged circulation in the plasma. The N-glycans in its structure protect it from the action of proteases and in the modulation of its receptor binding affinity. Mutations in EPO gene can cause microvascular complications in type II diabetic subjects and erythrocytosis. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bunn, HF (2013). Cold Spring Harb. Perspect. Med. 3(3); a011619).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service