Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408421

Sigma-Aldrich

Anti-ADAMTS18 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADAMTS21

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$4,261.00

R$4,261.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
50 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Estimated to ship onMay 30, 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~135.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by gene ADAMTS16, another family member. It is thought to function as a tumor suppressor. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. (provided by RefSeq)

Immunogen

ADAMTS18 (AAH63283.1, 1 a.a. ~ 1221 a.a) full-length human protein.

Sequence
MECALLLACAFPAAGSGPPRGLAGLGRVAKALQLCCLCCASVAAALASDSSSGASGLNDDYVFVTPVEVDSAGSYISHDILHNGRKKRSAQNARSSLHYRFSAFGQELHLELKPSAILSSHFIVQVLGKDGASETQKPEVQQCFYQGFIRNDSSSSVAVSTCAGLSGLIRTRKNEFLISPLPQLLAQEHNHSSPAGHHPHVLYKRTAEEKIQRYRGYPGSGRNYPGYSPSHIPHASQSRETEYHHRRLQKQHFCGRRKKYAPKPPTEDTYLRFDEYGSSGRPRRSAGKSQKGLNVETLVVADKKMVEKHGKGNVTTYILTVMNMVSGLFKDGTIGSDINVVVVSLILLEQEPGGLLINHHADQSLNSFCQWQSALIGKNGKRHDHAILLTGFDICSWKNEPCDTLGFAPISGMCSKYRSCTINEDTGLGLAFTIAHESGHNFGMIHDGEGNPCRKAEGNIMSPTLTGNNGVFSWSSCSRQYLKKFLSTPQAGCLVDEPKQAGQYKYPDKLPGQIYDADTQCKWQFGAKAKLCSLGFVKDICKSLWCHRVGHRCETKFMPAAEGTVCGLSMWCRQGQCVKFGELGPRPIHGQWSAWSKWSECSRTCGGGVKFQERHCNNPKPQYGGIFCPGSSRIYQLCNINPCNENSLDFRAQQCAEYNSKPFRGWFYQWKPYTKVEEEDRCKLYCKAENFEFFFAMSGKVKDGTPCSPNKNDVCIDGVCELVGCDHELGSKAVSDACGVCKGDNSTCKFYKGLYLNQHKANEYYPVVIIPAGARSIEIQELQVSSSYLAVRSLSQKYYLTGGWSIDWPGEFPFAGTTFEYQRSFNRPERLYAPGPTNETLVFEILMQGKNPGIAWKYALPKVMNGTPPATKRPAYTWSIVQSECSVSCGGGYINVKAICLRDQNTQVNSSFCSAKTKPVTEPKICNAFSCPAYWMPGEWSTCSKACAGGQQSRKIQCVQKKPFQKEEAVLHSLCPVSTPTQVQACNSHACPPQWSLGPWSQCSKTCGRGVRKRELLCKGSAAETLPESQCTSLPRPELQEGCVLGRCPKNSRLQWVASSWSECSATCGLGVRKREMKCSEKGFQGKLITFPERRCRNIKKPNLDLEETCNRRACPAHPVYNMVAGWYSLPWQQCTVTCGGGVQTRSVHCVQQGRPSSSCLLHQKPPVLRACNTNFCPAPEKREDPSCVDFFNWCHLVPQHGVCNHKFYGKQCCKSCTRKI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service