Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405052

Sigma-Aldrich

Monoclonal Anti-ANAPC2, (C-terminal) antibody produced in mouse

clone 7F2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

APC2, RP11-350O14.5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,261.00

R$4,261.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.88 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANAPC2(29882)

General description

A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits. The composition of the APC is highly conserved in organisms from yeast to humans. The product of this gene is a component of the complex and shares sequence similarity with a recently identified family of proteins called cullins, which may also be involved in ubiquitin-mediated degradation. (provided by RefSeq)

Immunogen

ANAPC2 (NP_037498, 716 a.a. ~ 822 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IEEERPQDRDNMVLIDSDDESDSGMASQADQKEEELLLFWTYIQAMLTNLESLSLDRIYNMLRMFVVTGPALAEIDLQELQGYLQKKVRDQQLVYSAGVYRLPKNCS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service