Skip to Content
Merck
All Photos(1)

Key Documents

S0755

Sigma-Aldrich

Dimethyl succinate

Synonym(s):

Succinic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH2COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein:
956776
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

0.3 mmHg ( 20 °C)

autoignition temp.

689 °F

expl. lim.

8.5 %

refractive index

n20/D 1.419 (lit.)

bp

200 °C (lit.)

mp

16-19 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Hamada et al.
Bioorganic & medicinal chemistry letters, 19(10), 2852-2855 (2009-04-11)
N-Oxalylglycine (NOG) derivatives were synthesized, and their inhibitory effect on histone lysine demethylase activity was evaluated. NOG and compound 1 inhibited histone lysine demethylases JMJD2A, 2C and 2D in enzyme assays, and their dimethyl ester prodrugs DMOG and 21 exerted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service