Merck
  • Synthesis and activity of N-oxalylglycine and its derivatives as Jumonji C-domain-containing histone lysine demethylase inhibitors.
Synthesis and activity of N-oxalylglycine and its derivatives as Jumonji C-domain-containing histone lysine demethylase inhibitors.
Bioorganic & medicinal chemistry letters (2009-04-11)
Shohei Hamada, Tae-Dong Kim, Takayoshi Suzuki, Yukihiro Itoh, Hiroki Tsumoto, Hidehiko Nakagawa, Ralf Janknecht, Naoki Miyata
ABSTRACT
N-Oxalylglycine (NOG) derivatives were synthesized, and their inhibitory effect on histone lysine demethylase activity was evaluated. NOG and compound 1 inhibited histone lysine demethylases JMJD2A, 2C and 2D in enzyme assays, and their dimethyl ester prodrugs DMOG and 21 exerted histone lysine methylating activity in cellular assays.
Sigma-Aldrich
Dimethyl succinate, 98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl bromoacetate, 98%
Pricing and availability is not currently available.