Skip to Content
Merck
All Photos(1)

Key Documents

RAB0726

Sigma-Aldrich

Human proNPPB / Natriuretic Peptides B ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
R$3,893.00

R$3,893.00


Estimated to ship onApril 08, 2025



Select a Size

Change View
96 WELLS
R$3,893.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

R$3,893.00


Estimated to ship onApril 08, 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.14 ng/mL
standard curve range: 0.137-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPPB(4879)

General description

The proBNP ELISA kit provides for the Quantitative measurement of proBNP in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirk J Lok et al.
European journal of heart failure, 16(9), 958-966 (2014-08-02)
A-type and B-type natriuretic peptides are established markers in chronic heart failure (HF). C-type natriuretic peptide (CNP) belongs to the same peptide family, but is predominantly localized in the endothelium. The prognostic role of CNP in heart failure has not
Abdoulaye Diané et al.
The Journal of endocrinology, 222(3), 327-339 (2014-07-25)
Pituitary adenylate cyclase-activating polypeptide (PACAP) is a widely distributed neuropeptide that acts as a neurotransmitter, neuromodulator, neurotropic factor, neuroprotectant, secretagogue, and neurohormone. Owing to its pleiotropic biological actions, knockout of Pacap (Adcyap1) has been shown to induce several abnormalities in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service